Learn More
Aviva Systems Biology Corp ZNF8 Antibody - N-terminal region (ARP39480_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF8. Alternate names: HF.18, Zfp128ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-57051