Learn More
Aviva Systems Biology Corp ZNF800 Antibody - N-terminal region (ARP32203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF800. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF800. Alternate names: FLJ43301, MGC130032, MGC130033ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54486