Learn More
Aviva Systems Biology Corp ZNF81 Antibody - middle region (ARP31985_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31985_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF81. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF81. Alternate names: HFZ20, MRX45, dJ54B20.6ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54405