Learn More
Aviva Systems Biology Corp ZNF83 Antibody - N-terminal region (ARP35952_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35952_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF83. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF83. Alternate names: FLJ11015, FLJ14876, FLJ30097, FLJ90585, HPF1, MGC33853, ZNF816BApplicationWB. Species reactivity: Human
Catalog No. 50-014-55942