Learn More
Aviva Systems Biology Corp ZNF883 Antibody - N-terminal region (ARP50949_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50949_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF883. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF883. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53844