Learn More
Aviva Systems Biology Corp ZNF91 Antibody - N-terminal region (ARP47574_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47574_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF91. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF91. Alternate names: HPF7, HTF10ApplicationWB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-014-59592