Learn More
Aviva Systems Biology Corp ZNF92 Antibody - C-terminal region (ARP39923_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39923_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF92. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF92. Alternate names: TF12, HPF12, HTF12ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57234