Learn More
Aviva Systems Biology Corp ZNFN1A1 Antibody - middle region (ARP32757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNFN1A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNFN1A1. Alternate names: IK1, LYF1, hIk-1, IKAROS, PRO0758, ZNFN1A1, Hs.54452ApplicationIHC, WB. Species reactivity: Dog, Human
Catalog No. 50-014-52873