Learn More
Cayman Chemical zngbr offIcnals 6Shogaol 10mg
SDP
Catalog No. 502952680
Encompass_Preferred
A minor, bioactive component of ginger with anti-inflammatory and anticancer properties; induces apoptosis in cancer cells through the production of reactive oxygen species and the activation of caspase, impairs tubulin polymerization, downregulates NF-κB signaling, inhibits the expression of inflammatory mediators, suppresses Toll-like receptor signaling, and enhances antioxidant defense mechanisms by inducing Nrf2/ARE-mediated gene expression
Catalog No. 50-295-2680 Supplier Cayman Chemical Supplier No. 1190110 MG
A minor, bioactive component of ginger with anti-inflammatory and anticancer properties; induces apoptosis in cancer cells through the production of reactive oxygen species and the activation of caspase, impairs tubulin polymerization, downregulates NF-κB signaling, inhibits the expression of inflammatory mediators, suppresses Toll-like receptor signaling, and enhances antioxidant defense mechanisms by inducing Nrf2/ARE-mediated gene expression