Learn More
Cayman Chemical 10-Shogaol, Purity: ≥98%, CAS: 36752-54-2, 10 mg
SDP
Catalog No. 502912003
Encompass_Preferred
A phenol with diverse biological activities; selectively inhibits COX-2 over COX-1 (IC50s = 7.5 and >50 µM, respectively); induces mortality in A. cantonensis larvae at 100 and 200 µM; scavenges DPPH radicals in a cell-free assay at 100 µM; increases proliferation of isolated human keratinocytes and fibroblasts and migration of isolated human keratinocytes in a wound healing assay at 2 µM
Catalog No. 50-291-2003 Supplier Cayman Chemical Supplier No. 3617110 MG
A phenol with diverse biological activities; selectively inhibits COX-2 over COX-1 (IC50s = 7.5 and >50 µM, respectively); induces mortality in A. cantonensis larvae at 100 and 200 µM; scavenges DPPH radicals in a cell-free assay at 100 µM; increases proliferation of isolated human keratinocytes and fibroblasts and migration of isolated human keratinocytes in a wound healing assay at 2 µM