Learn More
Cayman Chemical ZngIbrosIde R1 5mg
SDP
Catalog No. 502947561
Encompass_Preferred
A triterpene saponin and an active metabolite of ginsenoside Ro; cytotoxic to B16/F10 murine melanoma cells (IC50 = 24.52 UG/ml); inhibits tube formation by HUVECs at 40 UG/ml; reduces tumor growth in a B16/F10 murine melanoma model at 25 mg/kg
Catalog No. 50-294-7561 Supplier Cayman Chemical Supplier No. 341305 MG
A triterpene saponin and an active metabolite of ginsenoside Ro; cytotoxic to B16/F10 murine melanoma cells (IC50 = 24.52 UG/ml); inhibits tube formation by HUVECs at 40 UG/ml; reduces tumor growth in a B16/F10 murine melanoma model at 25 mg/kg