Learn More
Aviva Systems Biology Corp ZNHIT1 Antibody - N-terminal region (ARP50537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNHIT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNHIT1. Alternate names: CG1I, ZNFN4A1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60371