Learn More
FIREBIRD OPTICS Zinc Selenide (ZnSe) Circular Optical Window- 49x6mm
SDP
Catalog No. NC1953414
Encompass_Preferred
Zinc Selenide (ZnSe) Circular Optical Window- 49x6mm• Wavelength range: 1-14µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.13mm •Thickness tolerance: ±0.13mm
Catalog No. NC1953414 Supplier FIREBIRD OPTICS Supplier No. ZNSE49X6
Zinc Selenide (ZnSe) Circular Optical Window- 49x6mm• Wavelength range: 1-14µm •Surface quality: 60/40 • Clear aperture: 90% •Diameter tolerance: ±0.13mm •Thickness tolerance: ±0.13mm