Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zobell’s Solution, Reagents

Catalog No. 01333367
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-367 4 L
01-333-368 1 L
01-333-366 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-333-367 Supplier Reagents Holdings Llc Supplier No. CZ0001004B
Only null left
Add to Cart
Add to Cart

Zobell’s Solution, Reagent, use for Redox. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Zobell’s Solution
Packaging Opaque HDPE
CAS 7732-18-5, 7447-40-7, 14459-95-1
Specific Gravity 1.0048
Physical Form Liquid
Technique Redox
Color Yellow/Green
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Standard
Solubility Completely miscible
Quantity 4 L
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.