Learn More
Med Vet International Zoetis Advocin, 180mg/mL, 100mL, Generic Of Danofloxacin
Catalog No. 502827964
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
100ml Advocin (danofloxacin injection) from Zoetis is a sterile antimicrobial injectable solution that features the shortest withdrawal time of major antimicrobial brnads making it perfect for treating bovine respiratory disease (BRD) late in the feeding period. This pharmaceutical is fast-acting for BRD Therapy, has a 4 day meat withdrawal, and offers flexibility to treat animals during the entire feeding period.
Catalog No. 50-282-7964 Supplier Med Vet International Supplier No. RXADVOCIN
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
100ml Advocin (danofloxacin injection) from Zoetis is a sterile antimicrobial injectable solution that features the shortest withdrawal time of major antimicrobial brnads making it perfect for treating bovine respiratory disease (BRD) late in the feeding period. This pharmaceutical is fast-acting for BRD Therapy, has a 4 day meat withdrawal, and offers flexibility to treat animals during the entire feeding period.