Learn More
Med Vet International Zoetis Albon Tablets, 250mg, 500ct.
Catalog No. 502827879
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
250mg Albon (sulfadimethoxine) from Zoetis effectively treats a broad range of bacterial infections commonly found in dogs and cats. This product is indicated for use in dogs and cats with respiratory, genitourinary tract, and soft-tissue infections.
Catalog No. 50-282-7879 Supplier Med Vet International Supplier No. RXALBON250500
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
250mg Albon (sulfadimethoxine) from Zoetis effectively treats a broad range of bacterial infections commonly found in dogs and cats. This product is indicated for use in dogs and cats with respiratory, genitourinary tract, and soft-tissue infections.