Learn More
Med Vet International Zoetis Amoxi-Drops, 15mL, Vet Label
Catalog No. 502828602
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Amoxi-Drop (amoxicillin 50 mg/mL) is a semi-synthetic antibiotic with a broad spectrum of activity. It provides bactericidal activity against a wide range of common Gram-positive and Gram-negative pathogens.
Catalog No. 50-282-8602 Supplier Med Vet International Supplier No. RXAMOXIDR15
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Amoxi-Drop (amoxicillin 50 mg/mL) is a semi-synthetic antibiotic with a broad spectrum of activity. It provides bactericidal activity against a wide range of common Gram-positive and Gram-negative pathogens.