Learn More
Med Vet International Zoetis Covenia Injection, 10mL
Catalog No. 502828301
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dogs Convenia is indicated for the treatment of skin infections (secondary superficial pyoderma, abscesses, and wounds) in dogs caused by susceptible strains of Staphylococcus intermedius and Streptococcus canis (Group G). See package insert for complete product details. Cats Convenia is indicated for the treatment of skin infections (wounds and abscesses) in cats caused by susceptible strains of Pasteurella multocida. See package insert for complete product details. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. *Next day air available with additional charges*
Catalog No. 50-282-8301 Supplier Med Vet International Supplier No. RXCONVENIA
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dogs Convenia is indicated for the treatment of skin infections (secondary superficial pyoderma, abscesses, and wounds) in dogs caused by susceptible strains of Staphylococcus intermedius and Streptococcus canis (Group G). See package insert for complete product details. Cats Convenia is indicated for the treatment of skin infections (wounds and abscesses) in cats caused by susceptible strains of Pasteurella multocida. See package insert for complete product details. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. *Next day air available with additional charges*