Learn More
Med Vet International Zoetis Lutalyse, 5mg/mL, 100mL
Catalog No. 502828282
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dinoprost tromethamine sterile solution, used as a luteolytic agent in cattle, parturition induction in swine, and luteolytic effect in mares.
Catalog No. 50-282-8282 Supplier Med Vet International Supplier No. RXLUTALYSE100
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Dinoprost tromethamine sterile solution, used as a luteolytic agent in cattle, parturition induction in swine, and luteolytic effect in mares.