Learn More
Med Vet International Zoetis Simplicef Tablets, 200mg, 100ct.
Catalog No. 502828892
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Simplicef (cefpodoxime proxetil) tablets are indicated for the treatment of skin infections (wounds and abscesses) in dogs caused by susceptible strains of Staphylococcus intermedius, Staphylococcus aureus, Streptococcus canis (group G, B hemolytic), Escherichia coli, Pasteurella multocida, and Proteus mirabilis.
Catalog No. 50-282-8892 Supplier Med Vet International Supplier No. RXSIMPLICEF200
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Simplicef (cefpodoxime proxetil) tablets are indicated for the treatment of skin infections (wounds and abscesses) in dogs caused by susceptible strains of Staphylococcus intermedius, Staphylococcus aureus, Streptococcus canis (group G, B hemolytic), Escherichia coli, Pasteurella multocida, and Proteus mirabilis.