Learn More
Med Vet International Zoetis Zeniquin Tablets For Dogs And Cats, 25mg, 100ct., Generic Marbofloxacin
Catalog No. 502828476
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Zeniquin (Marbofloxacin) Tablets are indicated for the treatment of skin and soft-tissue infections in dogs and cats and urinary tract infections in dogs with bacteria susceptible to marbofloxacin. Marbofloxacin is a synthetic broad-spectrum antibacterial agent from the fluoroquinolone class of chemotherapeutic agents. Convenient once-a-day dosing. Scored and film-coated tablets are available in 4 sizes, providing flexible dosing for small to giant-sized breeds. Antibiotic for cats and dogs. Federal law prohibits the extra-label use of this drug in food-producing animals. 100 tablets per bottle.
Catalog No. 50-282-8476 Supplier Med Vet International Supplier No. RXZEN25100
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Zeniquin (Marbofloxacin) Tablets are indicated for the treatment of skin and soft-tissue infections in dogs and cats and urinary tract infections in dogs with bacteria susceptible to marbofloxacin. Marbofloxacin is a synthetic broad-spectrum antibacterial agent from the fluoroquinolone class of chemotherapeutic agents. Convenient once-a-day dosing. Scored and film-coated tablets are available in 4 sizes, providing flexible dosing for small to giant-sized breeds. Antibiotic for cats and dogs. Federal law prohibits the extra-label use of this drug in food-producing animals. 100 tablets per bottle.