Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zofenopril-d5 | 00-00-0 | 99.9% | 434.58 g/mol | C22H18D5NO4S2 | 1 MG
SDP

Catalog No. 5000243326
Encompass_Preferred

Zofenopril-d5 is a deuterium-labeled analogue of zofenopril intended for research use as an internal standard and tracer in analytical and metabolic studies. The d5 label provides a predictable mass shift for accurate quantification by LC-MS, GC-MS, and NMR while preserving the chemical properties of the parent compound.

  • Deuterium labeling (d5) for a clear mass shift in mass spectrometry.
  • High purity (99.9%).
  • Molecular weight 434.58 g/mol.
  • Molecular formula C22H18D5NO4S2.
  • Pack size 1 mg suitable for analytical workflows.
  • Storage recommendation: -20°C for long-term stability.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-43326 Supplier Medchemexpress LLC Supplier No. HY108321S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zofenopril-d5 is a deuterium-labeled analogue of zofenopril intended for research use as an internal standard and tracer in analytical and metabolic studies. The d5 label provides a predictable mass shift for accurate quantification by LC-MS, GC-MS, and NMR while preserving the chemical properties of the parent compound.

  • Deuterium labeling (d5) for a clear mass shift in mass spectrometry.
  • High purity (99.9%).
  • Molecular weight 434.58 g/mol.
  • Molecular formula C22H18D5NO4S2.
  • Pack size 1 mg suitable for analytical workflows.
  • Storage recommendation: -20°C for long-term stability.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.