Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zofenopril-d5 | 00-00-0 | 99.9% | 434.58 g/mol | C22H18D5NO4S2 | 10 MG
SDP

Catalog No. 5000243325
Encompass_Preferred

Zofenopril-d5 is a deuterium-labeled analog of zofenopril, an angiotensin-converting enzyme (ACE) inhibitor. Supplied for research use only, it is intended as a stable isotope standard for analytical, metabolic, and pharmacokinetic studies, including mass spectrometry applications.

  • Deuterium-labeled (d5) analog for isotope studies.
  • High purity suitable for analytical applications (≈99.9%).
  • Molecular weight 434.58 g/mol.
  • Chemical formula C22H18D5NO4S2.
  • Target: angiotensin-converting enzyme (ACE).
  • Intended for research use only, not for human use.
  • Recommended storage: -20 °C (solid); -80 °C in solution for long-term storage.

Catalog No. 50-002-43325 Supplier Medchemexpress LLC Supplier No. HY108321S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zofenopril-d5 is a deuterium-labeled analog of zofenopril, an angiotensin-converting enzyme (ACE) inhibitor. Supplied for research use only, it is intended as a stable isotope standard for analytical, metabolic, and pharmacokinetic studies, including mass spectrometry applications.

  • Deuterium-labeled (d5) analog for isotope studies.
  • High purity suitable for analytical applications (≈99.9%).
  • Molecular weight 434.58 g/mol.
  • Chemical formula C22H18D5NO4S2.
  • Target: angiotensin-converting enzyme (ACE).
  • Intended for research use only, not for human use.
  • Recommended storage: -20 °C (solid); -80 °C in solution for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.