Learn More
Med Vet International Zoguard Plus For Dogs, 5-22 Lbs., 3/Pack.
Catalog No. 502828619
Encompass_Preferred
ZoGuard Plus for Dogs (Fipronil/(S)-Methoprene) is a once-a-month dose topical application that is indicated to kill fleas, ticks (deer ticks, brown dog ticks, American dog ticks, lone star ticks) and chewing lice. For use on dogs and puppies 8 weeks of age or older and weighing 5 pounds or greater. See packaging for complete product details.
Catalog No. 50-282-8619 Supplier Med Vet International Supplier No. PHZOGRD522
ZoGuard Plus for Dogs (Fipronil/(S)-Methoprene) is a once-a-month dose topical application that is indicated to kill fleas, ticks (deer ticks, brown dog ticks, American dog ticks, lone star ticks) and chewing lice. For use on dogs and puppies 8 weeks of age or older and weighing 5 pounds or greater. See packaging for complete product details.