Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMSHUTTLE INC ChemShuttle Zoldonrasib, KRAS G12D Inhibitor, 98% Purity, 1 g

Catalog No. NC3955408
Encompass

Zoldonrasib (RMC-9805) is a potent inhibitor specifically targeting the KRAS G12D mutation. This chemical compound is designed for research applications, featuring a molecular formula of C63H88F3N11O7 and a molecular weight of 1168.44.

  • Purity of 98%
  • CAS number 2922732-54-3
  • Storage at 2-8 degrees Celsius

Catalog No. NC3955408 Supplier CHEMSHUTTLE INC Supplier No. 2000751G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zoldonrasib (RMC-9805) is a potent inhibitor specifically targeting the KRAS G12D mutation. This chemical compound is designed for research applications, featuring a molecular formula of C63H88F3N11O7 and a molecular weight of 1168.44.

  • Purity of 98%
  • CAS number 2922732-54-3
  • Storage at 2-8 degrees Celsius

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.