Learn More
Enzo Life Sciences Zoledronic acid (1g). CAS: 165800-06-6
SDP
Supplier:  Enzo Life Sciences ALX430153G001
Encompass_Preferred
Inhibitor of osteoclastic bone resorption. Prevents osteoporosis and skeletal fractures, particularly in patients with multiple myeloma or prostate cancer. Also used to treat hypercalcemia. Inhibition of acid sphingomyelinase (aSMase) with an IC50 5µm. Alternative name: 1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid, CGP 42446. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in 0.1N NaOH. Sparingly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9239