Learn More
Apexbio Technology LLC Zoledronic Acid(Synonyms: Zoledronate, Zometa, Reclast, Aclasta, CGP-42446, Zoledronic Acid Hydrate, Zoledronic Acid Monohydrate), 25mg, CAS: 118072-93-8.
SDP
Supplier:  Apexbio Technology LLC A135225
Encompass_Preferred
Zoledronic acid (CAS 118072-93-8) is a nitrogen-containing bisphosphonate widely studied for its potent cellular effects It exerts anti-proliferative and pro-apoptotic activity in vitro primarily by activating protein kinase C signaling pathways Research demonstrates that zoledronic acid inhibits proliferation and induces apoptosis in multiple myeloma cell lines Additionally investigations suggest its inhibitory effects extend to other cell types including human fetal cells Due to these cellular mechanisms zoledronic acid is frequently utilized in biomedical research focused on cancer proliferation apoptosis and therapeutic interventions
Catalog No. 50-000-60807