Learn More
Selleck Chemical LLC Zoledronic Acid 25mg 118072-93-8 Zoledronate, CGP-4244
SDP
Supplier:  Selleck Chemical LLC S131425MG
Encompass_Preferred
Zoledronic acid (ZA, Zoledronate, CGP-4244), a potent osteoclast inhibitor, induces apoptosis in osteoclasts by inhibiting enzymes of the mevalonate pathway and preventing the isoprenylation of small GTP-binding proteins such as Ras and Rho. Zoledronic acid (ZA) also induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1135