Learn More
Medchemexpress LLC Zoledronic acid-d3 (zoledronate-d3) | 1134798-26-7 | 275.11 | 1 MG
SDP
Catalog No. 5000347985
Encompass_Preferred
Zoledronic acid-d3 (Zoledronate-d3) is the deuterium labeled Zoledronic acid. Zoledronic Acid (Zoledronate) is a third-generation bisphosphonate (BP), with potent anti-resorptive activity. Zoledronic Acid inhibits the differentiation and apoptosis of osteoclasts. Zoledronic Acid also has anti-cancer effects.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-47985 Supplier Medchemexpress LLC Supplier No. HY13777S11MG
Zoledronic acid-d3 (Zoledronate-d3) is the deuterium labeled Zoledronic acid. Zoledronic Acid (Zoledronate) is a third-generation bisphosphonate (BP), with potent anti-resorptive activity. Zoledronic Acid inhibits the differentiation and apoptosis of osteoclasts. Zoledronic Acid also has anti-cancer effects.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.