Learn More
Selleck Chemical LLC Zoledronic acid monohydrate S5244-25mg
SDP
Catalog No. 503112241
Encompass_Preferred
Zoledronic acid (Zoledronate CGP-4244) monohydrate a nitrogen-containing bisphosphonate is a potent osteoclast inhibitor which induces apoptosis in osteoclasts by inhibiting enzymes of the mevalonate pathway and preventing the isoprenylation of small GTP-binding proteins such as Ras and Rho
Catalog No. 50-311-2241 Supplier Selleck Chemical LLC Supplier No. S524425MG
Zoledronic acid (Zoledronate CGP-4244) monohydrate a nitrogen-containing bisphosphonate is a potent osteoclast inhibitor which induces apoptosis in osteoclasts by inhibiting enzymes of the mevalonate pathway and preventing the isoprenylation of small GTP-binding proteins such as Ras and Rho