Learn More
Cayman Chemical Zoledronic Acid (hydrate), Purity: ≥95%, CAS: 165800-06-6, 50 mg
SDP
Catalog No. 502968509
Encompass_Preferred
A third-generation heterocyclic nitrogen-containing bisphosphonate that inhibits the prenylation of GTPases critical to the signaling events related to osteoclast-mediated bone resorption; has a high affinity for hydroxyapatite (Ki = 3.47 μM) and binds directly to mineralized bone where it decreases bone resorption by inhibiting osteoclast proliferation and inducing osteoclast apoptosis
Catalog No. 50-296-8509 Supplier Cayman Chemical Supplier No. 1498450 MG
A third-generation heterocyclic nitrogen-containing bisphosphonate that inhibits the prenylation of GTPases critical to the signaling events related to osteoclast-mediated bone resorption; has a high affinity for hydroxyapatite (Ki = 3.47 μM) and binds directly to mineralized bone where it decreases bone resorption by inhibiting osteoclast proliferation and inducing osteoclast apoptosis