ZOLL Pedi-padz II electrodes are designed for use on children up to eight years of age or weighing less than 25 kg. These pads ensure proper defibrillator electrode usage, as compatible ZOLL defibrillators provide voice and text prompts when connected. They utilize specific algorithms for children to analyze heart rhythm and automatically reduce energy for pediatric-specific therapy.
- Designed for children up to 8 years or 25 kg
- Anatomical diagrams on package for proper pad placement
- Compatible defibrillators provide voice and text prompts upon connection
- Utilizes specific algorithms for child heart rhythm analysis
- Defibrillator automatically reduces energy for pediatric therapy
- Compatible with AED Plus and professional defibrillators
- Does not use resistors or hardware for energy reduction