Learn More
ZOLL™ Medical AED Plus™ Trainer2
Catalog No. 17478012 Shop All Zoll Medical Corporation Products
Change view
Click to view available options
Product Type:
AED Plus Trainer2 Unit
Catalog No. Product Type
17-478-012 AED Plus Trainer2 Unit
1 options
Catalog No. 17-478-012 Supplier ZOLL™ Medical Supplier No. 8008005001
Add to Cart
Add to Cart
Includes
AED Plus Trainer2 training AED, wireless remote controller, one set of AED Plus Training CPR-D-padz™, one set of AED Plus CPR-D trainer pad reusable gels, four D cell batteries, two AA cell batteries, operator's guide
For CPR and first aid instructors to teach students on saving life with AED Plus
  • Wireless remote to control multiple AED Plus Trainer2 units
  • Instructors can set up rescue scenario and download in seconds
  • Software easily switches from AHA to ERC 2010 guidelines
Order Info
Replacement gels and electrodes available separately (Cat. Nos. 17-478-13 and -14).
Specifications
Product Type AED Plus Trainer2 Unit
Includes AED Plus Trainer2 training AED, wireless remote controller, one set of AED Plus training CPR-D-padz™, one set of AED Plus CPR-D trainer pad reusable gels, four D cell batteries, two AA cell batteries, operator's guide
For Use With (Application) Allows students to experience what it's like to help save a life with the AED Plus