Learn More
Selleck Chemical LLC Zolmitriptan 10mg 139264-17-8
SDP
Catalog No. 506383
Encompass_Preferred
Zolmitriptan is a novel and highly selective 5-HT(1B/1D) receptor agonist, used in the treatment of acute migraines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-638-3 Supplier Selleck Chemical LLC Supplier No. S164910MG
Zolmitriptan is a novel and highly selective 5-HT(1B/1D) receptor agonist, used in the treatment of acute migraines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.