Learn More
Cayman Chemical ZolmItrIptn 50mg
SDP
Catalog No. 502973028
Encompass_Preferred
A triptan that acts as an agonist of the serotonin (5-HT) receptors 5-HT1B and 5-HT1D (Kis = 2.2 and 0.92 nM, respectively); inhibits cAMP formation in blood vessels, resulting in vessel constriction
Catalog No. 50-297-3028 Supplier Cayman Chemical Supplier No. 1587150 MG
A triptan that acts as an agonist of the serotonin (5-HT) receptors 5-HT1B and 5-HT1D (Kis = 2.2 and 0.92 nM, respectively); inhibits cAMP formation in blood vessels, resulting in vessel constriction