Learn More
Research Products International Corp Zone-Free Adhesive Film, Non-Sterile, 50 per Package
SDP
Catalog No. 50998788
Encompass
This unique double-layer film consists of a clear, easily pierceable, adhesive-free polypropylene film bonded to a 96-well diecut adhesive polypropylene sublayer. This combination provides an adhesive-free zone over each well while ensuring a secure seal around the rim and between wells. Designed for 96-well microplates where well-to-well contamination and crossover is of critical concern. The easy-piercing film allows for direct sample recovery with single and multichannel pipettors or robotic probes. The adhesive-free zone above each well prevents adhesive fouling of the pipet tip or probe, ultimately minimizing the amount of adhesive that comes into contact with the sample. Pierceable with multichannel pipettors or robotic probes. Zone-Free is a Trademark of Excel Scientifc, Inc.
Catalog No. 50-998-788 Supplier Research Products International Corp Supplier No. 202515
This unique double-layer film consists of a clear, easily pierceable, adhesive-free polypropylene film bonded to a 96-well diecut adhesive polypropylene sublayer. This combination provides an adhesive-free zone over each well while ensuring a secure seal around the rim and between wells. Designed for 96-well microplates where well-to-well contamination and crossover is of critical concern. The easy-piercing film allows for direct sample recovery with single and multichannel pipettors or robotic probes. The adhesive-free zone above each well prevents adhesive fouling of the pipet tip or probe, ultimately minimizing the amount of adhesive that comes into contact with the sample. Pierceable with multichannel pipettors or robotic probes. Zone-Free is a Trademark of Excel Scientifc, Inc.