Learn More
BURKLE INC Zone sampler Spiralus, Alu/PA, 78.74 in. (200 cm), 37.19 oz. (1100 ml) vol.
Catalog No. 5000025874
Encompass_Preferred
The Spiralus zone sampler guarantees representative all-layer samples thanks to its spiral-shaped sample slots. The Spiralus zone sampler takes a zone-accurate all-layer sample through all layers of the bulk material. The spiral-shaped opening of the sample slots, which open with a time delay from the bottom to the top, enables the Spiralus to take a layer-accurate and representative sample. The special design of the Spiralus prevents grains from being cut and thus ensures excellent reproduction of the stratification when sampling from coarse-grained bulk materials.
The low weight of the Spiralus makes it easy to handle and use in different environments. With open handle for easy emptying of the mixed sample.
Catalog No. 50-000-25874 Supplier BURKLE INC Supplier No. 53180200
The Spiralus zone sampler guarantees representative all-layer samples thanks to its spiral-shaped sample slots. The Spiralus zone sampler takes a zone-accurate all-layer sample through all layers of the bulk material. The spiral-shaped opening of the sample slots, which open with a time delay from the bottom to the top, enables the Spiralus to take a layer-accurate and representative sample. The special design of the Spiralus prevents grains from being cut and thus ensures excellent reproduction of the stratification when sampling from coarse-grained bulk materials.
The low weight of the Spiralus makes it easy to handle and use in different environments. With open handle for easy emptying of the mixed sample.