Learn More
BURKLE INC Zone sampler Spiralus, Alu/PA, 57.09 in. (145 cm), 26.04 oz. (770 ml) volume
Catalog No. 5000025687
Encompass_Preferred
The Spiralus zone sampler guarantees representative all-layer samples thanks to its spiral-shaped sample slots. The Spiralus zone sampler takes a zone-accurate all-layer sample through all layers of the bulk material. The spiral-shaped opening of the sample slots, which open with a time delay from the bottom to the top, enables the Spiralus to take a layer-accurate and representative sample. The special design of the Spiralus prevents grains from being cut and thus ensures excellent reproduction of the stratification when sampling from coarse-grained bulk materials.
The low weight of the Spiralus makes it easy to handle and use in different environments. With open handle for easy emptying of the mixed sample.
Catalog No. 50-000-25687 Supplier BURKLE INC Supplier No. 53180145
The Spiralus zone sampler guarantees representative all-layer samples thanks to its spiral-shaped sample slots. The Spiralus zone sampler takes a zone-accurate all-layer sample through all layers of the bulk material. The spiral-shaped opening of the sample slots, which open with a time delay from the bottom to the top, enables the Spiralus to take a layer-accurate and representative sample. The special design of the Spiralus prevents grains from being cut and thus ensures excellent reproduction of the stratification when sampling from coarse-grained bulk materials.
The low weight of the Spiralus makes it easy to handle and use in different environments. With open handle for easy emptying of the mixed sample.