Learn More
Medchemexpress LLC HY-105064D 5mg Medchemexpress, Zoniporide (hydrochloride hydrate) CAS:863406-85-3 Purity:>98%
SDP
Catalog No. 501968247
Encompass_Preferred
Medchemexpress, HY-105064D 5mg Zoniporide (HCl hydrate) CAS:863406-85-3 Zoniporide (CP-597396) hydrochloride hydrate is a potent and selective inhibitor of sodium-hydrogen exchanger type 1 (NHE-1). Zoniporide hydrochloride hydrate inhibits human NHE-1 (IC50=14 nM), and has >150-fold selectivity versus other NHE isoforms. Zoniporide hydrochloride hydrate potently inhibits ex vivo NHE-1-dependent swelling of human platelets (IC50=59 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8247 Supplier Medchemexpress LLC Supplier No. HY105064D5MG
Medchemexpress, HY-105064D 5mg Zoniporide (HCl hydrate) CAS:863406-85-3 Zoniporide (CP-597396) hydrochloride hydrate is a potent and selective inhibitor of sodium-hydrogen exchanger type 1 (NHE-1). Zoniporide hydrochloride hydrate inhibits human NHE-1 (IC50=14 nM), and has >150-fold selectivity versus other NHE isoforms. Zoniporide hydrochloride hydrate potently inhibits ex vivo NHE-1-dependent swelling of human platelets (IC50=59 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.