Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zoniporide hydrochloride hydrate | 863406-85-3 | 99.4% | 374.82 g/mol | C17H19ClN6O2 | 25 MG
SDP

Catalog No. 5000243195
Encompass_Preferred

Zoniporide hydrochloride hydrate is the hydrochloride hydrate salt of zoniporide, a potent and selective inhibitor of the sodium-hydrogen exchanger isoform 1 (NHE-1) used for research applications. It is provided as a high-purity solid suitable for in vitro and ex vivo studies of NHE-1 function and related cardiovascular and platelet biology.

  • Potent and selective NHE-1 inhibition (human IC50 ~14 nM).
  • Inhibits ex vivo NHE-1-dependent platelet swelling (IC50 ~59 nM).
  • High purity (>99% by HPLC).
  • Available as the hydrochloride hydrate salt for improved handling and stability.
  • Molecular weight 374.82 g/mol; molecular formula C17H19ClN6O2.
  • Supplied in small research quantities, e.g., 25 MG.

Catalog No. 50-002-43195 Supplier Medchemexpress LLC Supplier No. HY105064D25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zoniporide hydrochloride hydrate is the hydrochloride hydrate salt of zoniporide, a potent and selective inhibitor of the sodium-hydrogen exchanger isoform 1 (NHE-1) used for research applications. It is provided as a high-purity solid suitable for in vitro and ex vivo studies of NHE-1 function and related cardiovascular and platelet biology.

  • Potent and selective NHE-1 inhibition (human IC50 ~14 nM).
  • Inhibits ex vivo NHE-1-dependent platelet swelling (IC50 ~59 nM).
  • High purity (>99% by HPLC).
  • Available as the hydrochloride hydrate salt for improved handling and stability.
  • Molecular weight 374.82 g/mol; molecular formula C17H19ClN6O2.
  • Supplied in small research quantities, e.g., 25 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.