Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zoniporide hydrochloride hydrate | 863406-85-3 | MFCD04113213 | 99.4% | 374.82 g/mol | C17H19ClN6O2 | 1 ML
SDP

Catalog No. 5000244220
Encompass_Preferred

Zoniporide hydrochloride hydrate is the hydrochloride hydrate salt of zoniporide (CP-597396), a potent and selective inhibitor of the sodium-hydrogen exchanger type 1 (NHE-1). Supplied as a ready-to-use solution, it is intended for research applications targeting membrane transporter and ion-channel pathways.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Potent, selective NHE-1 inhibitor; reported IC50 ≈ 14 nM.
  • High purity, typically ≥99.4% (HPLC).
  • Molecular formula C17H19ClN6O2; molecular weight 374.82 g/mol.
  • Soluble in DMSO (~20 mg/mL); may require warming or sonication.
  • Store sealed, away from moisture; in solvent store -80°C for long term, -20°C for short term.

Catalog No. 50-002-44220 Supplier Medchemexpress LLC Supplier No. HY105064D10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zoniporide hydrochloride hydrate is the hydrochloride hydrate salt of zoniporide (CP-597396), a potent and selective inhibitor of the sodium-hydrogen exchanger type 1 (NHE-1). Supplied as a ready-to-use solution, it is intended for research applications targeting membrane transporter and ion-channel pathways.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Potent, selective NHE-1 inhibitor; reported IC50 ≈ 14 nM.
  • High purity, typically ≥99.4% (HPLC).
  • Molecular formula C17H19ClN6O2; molecular weight 374.82 g/mol.
  • Soluble in DMSO (~20 mg/mL); may require warming or sonication.
  • Store sealed, away from moisture; in solvent store -80°C for long term, -20°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.