Learn More
Medchemexpress LLC Zoniporide hydrochloride hydrate | 863406-85-3 | MFCD04113213 | 99.4% | 374.82 g·mol⁻¹ | C17H19ClN6O2 | 50 MG
SDP
Catalog No. 5000243116
Encompass_Preferred
Zoniporide hydrochloride hydrate is the hydrated hydrochloride salt of zoniporide, a potent and selective inhibitor of the sodium-hydrogen exchanger type 1 (NHE-1). Supplied as a white to off-white solid for research use, the hydrated salt form improves aqueous solubility and is intended for in vitro and preclinical studies.
  • Potent NHE-1 inhibitor (IC50 ≈ 14 nM).
  • Greater than 150-fold selectivity over other NHE isoforms.
  • High aqueous solubility (H2O ≥ 100 mg/mL).
  • High purity (≈ 99.4%).
  • White to off-white solid appearance.
  • Stable when stored sealed and protected from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Available as a 50 mg research pack.
Catalog No. 50-002-43116 Supplier Medchemexpress LLC Supplier No. HY105064D50MG
Zoniporide hydrochloride hydrate is the hydrated hydrochloride salt of zoniporide, a potent and selective inhibitor of the sodium-hydrogen exchanger type 1 (NHE-1). Supplied as a white to off-white solid for research use, the hydrated salt form improves aqueous solubility and is intended for in vitro and preclinical studies.
  • Potent NHE-1 inhibitor (IC50 ≈ 14 nM).
  • Greater than 150-fold selectivity over other NHE isoforms.
  • High aqueous solubility (H2O ≥ 100 mg/mL).
  • High purity (≈ 99.4%).
  • White to off-white solid appearance.
  • Stable when stored sealed and protected from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • Available as a 50 mg research pack.