Learn More
Medchemexpress LLC Zoniporide hydrochloride hydrate | 863406-85-3 | MFCD32004449 | 99.4% | 374.82 g·mol⁻¹ | C17H19ClN6O2 | 10 MG
SDP
Catalog No. 5000243593
Encompass_Preferred
Zoniporide hydrochloride hydrate is the hydrochloride hydrate salt of zoniporide, a potent and selective inhibitor of the sodium-hydrogen exchanger isoform 1 (NHE-1). It inhibits human NHE-1 with an IC50 of 14 nM and shows strong selectivity over other NHE isoforms, making it suitable for in vitro research on NHE-1-mediated pathways. Supplied as a high-purity solid for laboratory research.
  • Potent NHE-1 inhibitor with an IC50 of 14 nM.
  • Greater than 150-fold selectivity versus other NHE isoforms.
  • High purity suitable for analytical and biological assays (purity reported 99.37%).
  • Molecular weight 374.82 g·mol⁻¹.
  • Available in small milligram pack sizes for research use.
Catalog No. 50-002-43593 Supplier Medchemexpress LLC Supplier No. HY105064D10MG
Zoniporide hydrochloride hydrate is the hydrochloride hydrate salt of zoniporide, a potent and selective inhibitor of the sodium-hydrogen exchanger isoform 1 (NHE-1). It inhibits human NHE-1 with an IC50 of 14 nM and shows strong selectivity over other NHE isoforms, making it suitable for in vitro research on NHE-1-mediated pathways. Supplied as a high-purity solid for laboratory research.
  • Potent NHE-1 inhibitor with an IC50 of 14 nM.
  • Greater than 150-fold selectivity versus other NHE isoforms.
  • High purity suitable for analytical and biological assays (purity reported 99.37%).
  • Molecular weight 374.82 g·mol⁻¹.
  • Available in small milligram pack sizes for research use.