Learn More
Cayman Chemical Zoniporide (hydrochloride), Purity: ≥98%, CAS: 241800-97-5, 10 mg
SDP
Catalog No. 502968992
Encompass_Preferred
Selectively inhibits human NHE-1 with an IC50 value of 14 nM; reduces infarct size in a rabbit myocardial ischemia-reperfusion model without adversely affecting hemodynamics or cardiac function
Catalog No. 50-296-8992 Supplier Cayman Chemical Supplier No. 1600410 MG
Selectively inhibits human NHE-1 with an IC50 value of 14 nM; reduces infarct size in a rabbit myocardial ischemia-reperfusion model without adversely affecting hemodynamics or cardiac function