Learn More
Cayman Chemical Zoniporide (hydrochloride), Purity: ≥98%, CAS: 241800-97-5, 50 mg
SDP
Catalog No. 502968537
Encompass_Preferred
Selectively inhibits human NHE-1 with an IC50 value of 14 nM; reduces infarct size in a rabbit myocardial ischemia-reperfusion model without adversely affecting hemodynamics or cardiac function
Catalog No. 50-296-8537 Supplier Cayman Chemical Supplier No. 1600450 MG
Selectively inhibits human NHE-1 with an IC50 value of 14 nM; reduces infarct size in a rabbit myocardial ischemia-reperfusion model without adversely affecting hemodynamics or cardiac function