Learn More
Medchemexpress LLC Zonisamide-d4 | 1020720-04-0 | 99.4% | 1 MG
SDP
Catalog No. 5000450881
Encompass_Preferred
Zonisamide-d4 is a deuterium-labeled version of Zonisamide, which is an inhibitor of the zinc enzyme carbonic anhydrase (CA). It demonstrates antiepileptic activity and is utilized in research for epilepsy, seizures, and Parkinson's disease. Deuterium labeling has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
Catalog No. 50-004-50881 Supplier Medchemexpress LLC Supplier No. HYB0124S1MG
Zonisamide-d4 is a deuterium-labeled version of Zonisamide, which is an inhibitor of the zinc enzyme carbonic anhydrase (CA). It demonstrates antiepileptic activity and is utilized in research for epilepsy, seizures, and Parkinson's disease. Deuterium labeling has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.