Description
- Professional stereo microscope with a 6:1 zoom ratio
- Continuous magnification from 7.5X to 45X
- 110 mm working distance accommodates larger specimens
- Top and bottom LED illumination with selectable operating modes
- Heavy-duty all-metal frame with adjustable viewing head position
- Suitable for industrial, inspection, and educational applications
Specifications
Specifications
| Voltage | 110/220 V |
| Magnification Power | 0.75X to 4.5X |
| Objective Types | Achromatic |
| Product Type | Stereo Microscope |
| Head Style | Binocular |
| Lighting Type | LED |
| Eyepiece | WF 10X |