Learn More
Agilent Technologies ZORBAX ECLIPSE XDB-C18 4.6X150
Catalog No. NC2597283
Encompass
We’re currently building out the product information and will be adding additional content soon. Thank you for your patience as we continue to expand our selection and improve your ordering experience.
Catalog No. NC2597283 Supplier Agilent Technologies Supplier No. 993967902