Learn More
Neta Scientific ZORBAX RRHD 300-HILIC,2.1X100M
SDP
Supplier:  Neta Scientific 858750901
Encompass_Preferred
This item is manufactured by Agilent
ZORBAX RRHD 300-HILIC,2.1x100mm,1.8um
Catalog No. NC2189334