Learn More
Agilent Technologies ZORBAX RRHD STABLEBOND PHENYL
Catalog No. NC2392901
Encompass
ZORBAX RRHD StableBond Phenyl, 2.1 x 100 mm, 1.8 um, 1200 bar. 80A
Catalog No. NC2392901 Supplier Agilent Technologies Supplier No. 858700912
ZORBAX RRHD StableBond Phenyl, 2.1 x 100 mm, 1.8 um, 1200 bar. 80A